4,5-Dibromo-2-fluoroanisol ≥95%

Supplier: AOBChem USA

Warning

14874-1G 14874-5G 14874-25G
77341-658EA 98.88 USD
77341-658 77360-188 77372-152
4,5-Dibromo-2-fluoroanisol ≥95%
4,5-Dibromo-2-fluoroanisol

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
MW: 283.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527398
CAS Number: 861928-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR