2-Fluoro-3,6-diiodoanisole ≥97%

Supplier: AOBChem USA

Warning

14859-500MG 14859-1G 14859-5G
77323-982EA 311.06 USD
77323-982 77347-980 77372-150
2-Fluoro-3,6-diiodoanisole ≥97%
2-Fluoro-3,6-diiodoanisole

Precaución: for Research Use Only

Formula: C₇H₅FI₂O
Storage Temperature: Ambient
MDL Number: MFCD28791261
CAS Number: 861928-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR