3-Fluoro-1-(methoxymethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

14768-500MG 14768-1G 14768-5G 14768-25G
77323-978EA 119.48 USD
77323-978 77343-624 77360-178 77372-138
3-Fluoro-1-(methoxymethoxy)-2-methylbenzene ≥95%
3-Fluoro-1-(methoxymethoxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619361
CAS Number: 2179038-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR