4-(Quinolin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

147606-500MG 147606-1G 147606-5G 147606-25G
77323-976EA 223.51 USD
77323-976 77347-976 77360-176 77383-822
4-(Quinolin-3-yl)benzaldehyde ≥97%
4-(Quinolin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₁NO
MW: 233.265 g/mol
Storage Temperature: Ambient
CAS Number: 893735-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR