2-Fluoro-5-methyl-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

147325-500MG 147325-1G 147325-5G
77339-868EA 664.35 USD
77339-868 77347-970 77372-130
2-Fluoro-5-methyl-3-phenylpyridine ≥97%
2-Fluoro-5-methyl-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
CAS Number: 1214329-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR