4-(6-Methylpyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

147305-500MG 147305-1G 147305-5G
77323-970EA 316.21 USD
77323-970 77347-966 77372-128
4-(6-Methylpyridin-3-yl)benzaldehyde ≥97%
4-(6-Methylpyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.233 g/mol
Storage Temperature: Ambient
CAS Number: 1339711-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR