6-Methyl-5-phenylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

147292-500MG 147292-1G 147292-5G
77337-572EA 423.33 USD
77337-572 77347-964 77372-126
6-Methyl-5-phenylpyridin-2-amine ≥97%
6-Methyl-5-phenylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
Storage Temperature: Ambient
CAS Number: 84596-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR