6-Methyl-5-(4-(trifluoromethyl)phenyl)pyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

147286-500MG 147286-1G 147286-5G 147286-25G
77323-968EA 182.31 USD
77323-968 77347-106 77360-170 77383-816
6-Methyl-5-(4-(trifluoromethyl)phenyl)pyridin-2-amine ≥97%
6-Methyl-5-(4-(trifluoromethyl)phenyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₁F₃N₂
MW: 252.235 g/mol
Storage Temperature: Ambient
CAS Number: 1261733-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR