5-Methyl-[3,4'-bipyridin]-2-amine ≥97%

Supplier: AOBChem USA

Warning

147275-500MG 147275-1G 147275-5G
77323-966EA 309 USD
77323-966 77347-962 77372-124
5-Methyl-[3,4'-bipyridin]-2-amine ≥97%
5-Methyl-[3,4'-bipyridin]-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁N₃
MW: 185.225 g/mol
Storage Temperature: Ambient
CAS Number: 1214369-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR