3-(3-Fluorophenyl)-5-methylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

147273-500MG 147273-1G 147273-5G 147273-25G
77323-964EA 202.91 USD
77323-964 77347-960 77360-168 77383-814
3-(3-Fluorophenyl)-5-methylpyridin-2-amine ≥97%
3-(3-Fluorophenyl)-5-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₁FN₂
MW: 202.228 g/mol
Storage Temperature: Ambient
CAS Number: 1214349-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR