5-Methyl-3-(4-(trifluoromethyl)phenyl)pyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

147272-500MG 147272-1G 147272-5G 147272-25G
77323-962EA 202.91 USD
77323-962 77347-958 77360-166 77383-812
5-Methyl-3-(4-(trifluoromethyl)phenyl)pyridin-2-amine ≥97%
5-Methyl-3-(4-(trifluoromethyl)phenyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₁F₃N₂
MW: 252.235 g/mol
Storage Temperature: Ambient
CAS Number: 1261786-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR