2,2-Dichloro-1-(2,6-dibromophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

146690-500MG 146690-1G 146690-5G 146690-25G
77323-960EA 141.11 USD
77323-960 77344-962 77360-162 77383-810
2,2-Dichloro-1-(2,6-dibromophenyl)ethanol ≥95%
2,2-Dichloro-1-(2,6-dibromophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₆Br₂Cl₂O
Storage Temperature: Ambient
CAS Number: 2921693-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR