2-Bromo-1-butoxy-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

14563-500MG 14563-1G 14563-5G 14563-25G
77323-958EA 156.56 USD
77323-958 77345-596 77360-156 77383-808
2-Bromo-1-butoxy-4-methylbenzene ≥95%
2-Bromo-1-butoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156053
CAS Number: 57685-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR