2,6-dimethoxy-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

145619-500MG 145619-1G 145619-5G 145619-25G
77323-956EA 276.04 USD
77323-956 77347-954 77360-154 77383-806
2,6-dimethoxy-4-(trifluoromethyl)benzaldehyde ≥97%
2,6-dimethoxy-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
CAS Number: 1783617-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR