4-Bromo-2-ethyl-6-iodoaniline ≥95%

Supplier: AOBChem USA

Warning

145293-500MG 145293-1G 145293-5G
77338-392EA 484.1 USD
77338-392 77347-950 77372-114
4-Bromo-2-ethyl-6-iodoaniline ≥95%
4-Bromo-2-ethyl-6-iodoaniline

Precaución: for Research Use Only

Formula: C₈H₉BrIN
Storage Temperature: Ambient
CAS Number: 1520590-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR