N-(5-Bromopyridin-2-yl)isonicotinamide ≥97%

Supplier: AOBChem USA

Warning

145286-500MG 145286-1G 145286-5G 145286-25G
77323-952EA 161.71 USD
77323-952 77346-070 77360-150 77383-802
N-(5-Bromopyridin-2-yl)isonicotinamide ≥97%
N-(5-Bromopyridin-2-yl)isonicotinamide

Precaución: for Research Use Only

Formula: C₁₁H₈BrN₃O
MW: 278.1047 g/mol
Storage Temperature: Ambient
CAS Number: 310452-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR