4-Fluoro-2-(trifluoromethyl)anisole ≥95%

Supplier: AOBChem USA

Warning

14405-500MG 14405-1G 14405-5G 14405-25G
77323-948EA 150.38 USD
77323-948 77345-328 77360-142 77383-800
4-Fluoro-2-(trifluoromethyl)anisole ≥95%
4-Fluoro-2-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972882
CAS Number: 1214360-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR