3-Methoxy-4-thiomethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14323-500MG 14323-1G 14323-25G 14323-5G
77323-946EA 289.43 USD
77323-946 77347-944 77383-798 77372-098
3-Methoxy-4-thiomethylbenzoic acid ≥97%
3-Methoxy-4-thiomethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃S
MW: 198.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11103017
CAS Number: 95420-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR