2-Iodo-3-methoxybenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
14315-500MG
14315-1G
14315-5G
14315-25G
77323-944EA
181.28
USD
77323-944
77346-770
77360-138
77383-796
2-Iodo-3-methoxybenzoic acid ≥95%
2-Iodo-3-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ MW: 278.044 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18904957 CAS Number: 50627-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...