4-Bromo-3,5-dichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

14234-500MG 14234-1G 14234-5G 14234-25G
77323-940EA 154.5 USD
77323-940 77345-526 77360-136 77383-794
4-Bromo-3,5-dichlorotoluene ≥97%
4-Bromo-3,5-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447585
CAS Number: 19393-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR