5-Fluoro-3,3'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

142053-500MG 142053-1G 142053-5G
77338-588EA 501.61 USD
77338-588 77347-940 77372-092
5-Fluoro-3,3'-bipyridine ≥97%
5-Fluoro-3,3'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇FN₂
MW: 174.174 g/mol
Storage Temperature: Ambient
CAS Number: 1214360-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR