4-(5-Methylpyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

142014-500MG 142014-1G 142014-5G 142014-25G
77323-936EA 223.51 USD
77323-936 77347-938 77360-132 77383-792
4-(5-Methylpyridin-3-yl)benzaldehyde ≥97%
4-(5-Methylpyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.233 g/mol
Storage Temperature: Ambient
CAS Number: 1547915-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR