2-Bromo-1-chloro-4-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

14189-500MG 14189-1G 14189-5G 14189-25G
77323-934EA 166.86 USD
77323-934 77346-180 77360-130 77383-790
2-Bromo-1-chloro-4-ethoxybenzene ≥97%
2-Bromo-1-chloro-4-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD24153924
CAS Number: 1539442-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR