3-Ethoxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

140684-500MG 140684-1G 140684-5G 140684-25G
77323-928EA 243.08 USD
77323-928 77347-936 77360-120 77383-786
3-Ethoxy-4-(trifluoromethyl)benzoic acid ≥97%
3-Ethoxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
CAS Number: 911111-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR