2-(Benzyloxy)-4-bromo-1-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

140679-1G 140679-5G 140679-25G
77341-654EA 97.85 USD
77341-654 77360-118 77372-080
2-(Benzyloxy)-4-bromo-1-(trifluoromethyl)benzene ≥97%
2-(Benzyloxy)-4-bromo-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
CAS Number: 1314987-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR