4-Bromo-2-isopropoxy-1-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

140676-500MG 140676-1G 140676-5G 140676-25G
77323-926EA 146.26 USD
77323-926 77345-190 77360-116 77383-784
4-Bromo-2-isopropoxy-1-(trifluoromethyl)benzene ≥97%
4-Bromo-2-isopropoxy-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Ambient
CAS Number: 1369905-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR