3-(3-Bromophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14027-500MG 14027-1G 14027-5G 14027-25G
77323-922EA 142.14 USD
77323-922 77344-964 77360-112 77383-782
3-(3-Bromophenylcarbamoyl)-4-fluorophenylboronic acid ≥97%
3-(3-Bromophenylcarbamoyl)-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BBrFNO₃
MW: 337.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856000
CAS Number: 1451393-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR