1-(3,5-Dichlorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139889-500MG 139889-1G 139889-5G 139889-25G
77323-920EA 249.26 USD
77323-920 77347-934 77360-108 77383-780
1-(3,5-Dichlorophenyl)-2-methylpropan-1-ol ≥97%
1-(3,5-Dichlorophenyl)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O
MW: 219.108 g/mol
Storage Temperature: Ambient
CAS Number: 1341140-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR