1,2-Dibromo-4-ethoxy-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

138972-1G 138972-5G 138972-25G
77342-102EA 113.3 USD
77342-102 77360-102 77372-066
1,2-Dibromo-4-ethoxy-5-methoxybenzene ≥97%
1,2-Dibromo-4-ethoxy-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂O₂
Storage Temperature: Ambient
CAS Number: 2384462-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR