1-Bromo-5-fluoro-2-nitro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

138971-500MG 138971-1G 138971-5G 138971-25G
77323-910EA 151.41 USD
77323-910 77345-438 77360-100 77383-776
1-Bromo-5-fluoro-2-nitro-4-(trifluoromethyl)benzene ≥97%
1-Bromo-5-fluoro-2-nitro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrF₄NO₂
MW: 287.99 g/mol
Storage Temperature: Ambient
CAS Number: 1360438-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR