2-Chloro-2,2-difluoro-1-(2-fluoro-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138468-500MG 138468-1G 138468-5G
77337-598EA 424.36 USD
77337-598 77347-924 77372-064
2-Chloro-2,2-difluoro-1-(2-fluoro-4-methylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluoro-4-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
CAS Number: 1824274-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR