2-Chloro-2,2-difluoro-1-(4-fluoro-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

138464-500MG 138464-1G 138464-5G 138464-25G
77323-908EA 220.42 USD
77323-908 77347-922 77360-098 77383-774
2-Chloro-2,2-difluoro-1-(4-fluoro-2-methylphenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(4-fluoro-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
CAS Number: 1352205-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR