(5-Bromopyridin-2-yl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

13828-500MG 13828-1G 13828-5G 13828-25G
77323-906EA 265.74 USD
77323-906 77347-920 77360-096 77383-772
(5-Bromopyridin-2-yl)(4-methylpiperazin-1-yl)methanone ≥97%
(5-Bromopyridin-2-yl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN₃O
Storage Temperature: Ambient
MDL Number: MFCD17259448
CAS Number: 909712-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR