5-Bromo-N-(tert-butyl)picolinamide ≥97%

Supplier: AOBChem USA

Warning

13817-500MG 13817-1G 13817-5G 13817-25G
77323-904EA 187.46 USD
77323-904 77347-918 77360-094 77383-770
5-Bromo-N-(tert-butyl)picolinamide ≥97%
5-Bromo-N-(tert-butyl)picolinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD17259445
CAS Number: 647826-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR