7-Chloro-4-iodo-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
137887-500MG
137887-1G
137887-5G
77340-274EA
872.41
USD
77340-274
77347-914
77372-060
7-Chloro-4-iodo-1H-indazole ≥97%
7-Chloro-4-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄ClIN₂ MW: 278.478 g/mol Storage Temperature: Ambient |
CAS Number:
2921857-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...