2-Hydroxy-6-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

13748-500MG 13748-1G 13748-25G 13748-5G
77323-900EA 301.79 USD
77323-900 77347-912 77383-768 77372-056
2-Hydroxy-6-(trifluoromethoxy)benzoic acid ≥97%
2-Hydroxy-6-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₄
MW: 222.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397460
CAS Number: 851341-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR