3-Isopropoxy-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

135553-500MG 135553-1G 135553-5G
77339-280EA 571.65 USD
77339-280 77347-908 77372-052
3-Isopropoxy-2,4-dimethoxybenzoic acid ≥97%
3-Isopropoxy-2,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
Storage Temperature: Ambient
CAS Number: 2921712-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR