3-Isopropoxy-2,4-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

135548-500MG 135548-1G 135548-5G
77340-132EA 778.68 USD
77340-132 77347-906 77372-050
3-Isopropoxy-2,4-dimethoxybenzaldehyde ≥95%
3-Isopropoxy-2,4-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
Storage Temperature: Ambient
CAS Number: 101923-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR