(4-Chlorophenyl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

135261-500MG 135261-1G 135261-5G 135261-25G
77323-896EA 121.54 USD
77323-896 77343-898 77360-086 77372-044
(4-Chlorophenyl)(thiophen-2-yl)methanol ≥97%
(4-Chlorophenyl)(thiophen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₉ClOS
MW: 224.707 g/mol
Storage Temperature: Ambient
CAS Number: 40310-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR