4-Fluoro-2-(hydroxymethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

134651-1G 134651-5G 134651-25G
77341-652EA 97.85 USD
77341-652 77360-082 77372-040
4-Fluoro-2-(hydroxymethyl)phenol ≥97%
4-Fluoro-2-(hydroxymethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412237
CAS Number: 2357-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR