2-(Methylthio)-3-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(Methylthio)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
13459-500MG
13459-1G
13459-5G
13459-25G
77323-892EA
254.41
USD
77323-892
77347-902
77360-080
77383-764
2-(Methylthio)-3-pyridineboronic acid pinacol ester ≥97%
2-(Methylthio)-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₈BNO₂S MW: 251.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09037492 CAS Number: 1073354-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...