2-Bromo-6-methylbenzene-1,4-diol ≥97%

Supplier: AOBChem USA

Warning

134549-500MG 134549-1G 134549-5G 134549-25G
77323-890EA 120.51 USD
77323-890 77343-862 77360-078 77372-038
2-Bromo-6-methylbenzene-1,4-diol ≥97%
2-Bromo-6-methylbenzene-1,4-diol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
MW: 203.033 g/mol
Storage Temperature: Ambient
CAS Number: 50848-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR