4-Chloro-3-fluoro-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

134398-1G 134398-5G 134398-25G
77341-650EA 97.85 USD
77341-650 77360-076 77372-036
4-Chloro-3-fluoro-2-methylaniline ≥97%
4-Chloro-3-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇ClFN
Storage Temperature: Ambient
CAS Number: 1096113-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR