2-Bromo-1-chloro-4-ethynylbenzene ≥97%

Supplier: AOBChem USA

Warning

133328-500MG 133328-1G 133328-5G
77339-176EA 559.29 USD
77339-176 77347-898 77372-026
2-Bromo-1-chloro-4-ethynylbenzene ≥97%
2-Bromo-1-chloro-4-ethynylbenzene

Precaución: for Research Use Only

Formula: C₈H₄BrCl
Storage Temperature: Ambient
CAS Number: 1564565-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR