3-Acetyl-6-(trifluoromethyl)-2H-chromen-2-one ≥97%

Supplier: AOBChem USA

Warning

132758-500MG 132758-1G 132758-5G 132758-25G
77323-884EA 130.81 USD
77323-884 77344-322 77360-068 77372-024
3-Acetyl-6-(trifluoromethyl)-2H-chromen-2-one ≥97%
3-Acetyl-6-(trifluoromethyl)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₂H₇F₃O₃
MW: 256.1773 g/mol
Storage Temperature: Ambient
CAS Number: 2918958-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR