2-Chloro-3-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

13265-1G 13265-5G 13265-25G
77342-104EA 114.33 USD
77342-104 77360-064 77372-022
2-Chloro-3-hydroxybenzoic acid ≥95%
2-Chloro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD09264192
CAS Number: 51786-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR