7-Iodo-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

132582-500MG 132582-1G 132582-5G
77323-880EA 313.12 USD
77323-880 77347-894 77372-020
7-Iodo-1-methyl-1H-indazole ≥97%
7-Iodo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
Storage Temperature: Ambient
CAS Number: 1337879-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR