2-Chloro-5-hydroxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

132485-500MG 132485-1G 132485-5G 132485-25G
77323-878EA 128.75 USD
77323-878 77344-292 77360-062 77372-018
2-Chloro-5-hydroxyisonicotinic acid ≥95%
2-Chloro-5-hydroxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189124
CAS Number: 1060804-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR