3-Bromo-4-fluorophenyl methyl sulfone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-fluoro-4-(methylsulphonyl)benzene

131787-500MG 131787-1G 131787-5G 131787-25G
77323-876EA 191.58 USD
77323-876 77347-892 77360-060 77383-760
3-Bromo-4-fluorophenyl methyl sulfone ≥97%
3-Bromo-4-fluorophenyl methyl sulfone

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂S
MW: 253.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032287
CAS Number: 959961-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR