1-Bromo-4-fluoro-2-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

131706-500MG 131706-1G 131706-5G 131706-25G
77323-874EA 121.54 USD
77323-874 77343-974 77360-058 77372-016
1-Bromo-4-fluoro-2-(methylsulfonyl)benzene ≥97%
1-Bromo-4-fluoro-2-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂S
MW: 253.089 g/mol
Storage Temperature: Ambient
CAS Number: 628692-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR